Pump tubing, Viton®
Supplier: Heidolph Instruments GmbH & Co.KG
Certificates
About this item
Fluorocarbon rubber, thermoformed Viton B (67% fluorinated), opaque black.
- Low gas permeability
- Resistant to solvents and corrosives at high temperatures
Excellent acid resistance at high temperatures.
Temperature range: -30 to +205 °C
Certifications: GLP conform.
Frequently Bought Together