ContactUsLinkComponent
Disinfectant products, Stabimed® fresh
Disinfectant products, Stabimed® fresh
Catalog # BRAU19690
Supplier:  B. Braun
New Product
undefined
Disinfectant products, Stabimed® fresh
Catalog # BRAU19690
Supplier:  B. Braun
New Product
Specifications
  • Capacity:
    5 L
  • Description:
    Stabimed® fresh, for disinfection and cleaning of thermostable and thermolabile instruments
  • Seq:
    02
  • Pk:
    5 L
Specifications
About this item
Stabimed® for disinfection and purification of thermolabile and thermostable instruments. Aldehyde and phenol and QAC-free. Fast and gentle treatment of surgical and dental instruments, rigid and flexible endoscopes, anesthetic equipment, laboratory equipment and other thermolabile materials.
  • Active against bacteria, fungi, enveloped viruses, rotavirus, polyamo and adeno viruses and vaccinia virus
  • Highly economic
  • Used in ultrasonic bath
  • Exceptional cleaning power combined with optimal blood and secretions solvent power