Order Entry
Austria
ContactUsLinkComponent
Uridine-5'-triphosphoric acid trisodium salt (UTP-Na3) dihydrate
Uridine-5'-triphosphoric acid trisodium salt (UTP-Na3) dihydrate
Catalog # MOLE11038928-1G
Supplier:  Molekula
CAS Number:  
Uridine-5'-triphosphoric acid trisodium salt (UTP-Na3) dihydrate
Catalog # MOLE11038928-1G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :586.12 g/mol
Storage Temperature:Freezer
EINECS:243-347-5
CAS Number:116295-90-0
Formula:C₉H₁₂N₂Na₃O₁₅P₃·2H₂O