Uridine-5'-triphosphoric acid trisodium salt (UTP-Na3), Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:19817-92-6
:
Formula:C₉H₁₂N₂Na₃O₁₅P₃
MDL Number:MFCD00044310
:550.09 g/mol
Storage Temperature:Freezer
EINECS:243-347-5
Frequently Bought Together