Uridine-5'-triphosphoric acid trisodium salt (UTP-Na3), Millipore®
Catalog # 6701-50
Supplier: Merck Millipore (Calbiochem)
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:50 mg
- Seq:2370
Specifications
About this item
:
:550.09 g/mol
Storage Temperature:Freezer
EINECS:243-347-5
CAS Number:19817-92-6
Formula:C₉H₁₂N₂Na₃O₁₅P₃
MDL Number:MFCD00044310