AEBSF hydrochloride (4-(2-aminoethyl)benzenesulphonylfluoride hydrochloride)
Catalog # PIER78431
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:239.69 g/mol
Storage Temperature:Refrigerator
UN:3261
ADR:8,II
:GHS07
CAS Number:30827-99-7
Formula:C₈H₁₁ClFNO₂S
MDL Number:MFCD00132962