Order Entry
Austria
ContactUsLinkComponent
 
AEBSF hydrochloride (4-(2-aminoethyl)benzenesulphonylfluoride hydrochloride) ≥98%, Proteomics Grade
  M182-500MG
 :  
undefined
AEBSF hydrochloride (4-(2-aminoethyl)benzenesulphonylfluoride hydrochloride) ≥98%, Proteomics Grade
  M182-500MG
 :  

 

  • Pk:
    500 mg
  • Seq:
    2725

 

 

  • Protease:
    NONE
  • Purity (Titration):
    98 %
  • Solubility (0.02%, Water):
    PASS

 

 

 :
 :239.69 g/mol
 :Refrigerator
 :3261
 :8,II
 :GHS07
 :30827-99-7
 :C₈H₁₁ClFNO₂S
 :MFCD00132962