Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Enzymes
Bovine alpha-Chymotrypsin (from Pancreas)
Bovine alpha-Chymotrypsin (from Pancreas)
Bovine alpha-Chymotrypsin (from Pancreas)
SIALCHY5S-10VL
:
Merck
:
Bovine alpha-Chymotrypsin (from Pancreas)
SIALCHY5S-10VL
:
Merck
:
Pk:
10 Vial
CAS No.:
9004-07-3
Source:
Pancreas
Species:
Bovine
Storage conditions:
Freezer
MDL No.:
MFCD00130481
Enzyme name:
alpha-Chymotrypsin