N-Dodecyl-N,N-dimethyl-3-ammonio-1-propanesulphonate(sulphobetaine-12, SB-12)
Supplier: G-Biosciences
Certificates
About this item
CAS Number:14933-08-5
:
Formula:C₁₇H₃₇NO₃S
MDL Number:MFCD00036909
:335.55 g/mol
Storage Temperature:Ambient
EINECS:239-002-3
UN:3261
ADR:8,II
:GHS05
Frequently Bought Together