Order Entry
Austria
ContactUsLinkComponent
N-Decyl-N,N-dimethyl-3-ammonio-1-propanesulphonate(sulphobetaine-10, SB-10) ≥99% for biotechnology
 
N-Decyl-N,N-dimethyl-3-ammonio-1-propanesulphonate(sulphobetaine-10, SB-10) ≥99% for biotechnology
  J548-10G
 :  
undefined
 
N-Decyl-N,N-dimethyl-3-ammonio-1-propanesulphonate(sulphobetaine-10, SB-10) ≥99% for biotechnology
  J548-10G
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    10 g
  • Seq:
    3236

 

 

  • Appearance:
    White powder
  • Moisture (KF):
    = 2 %
  • Purity:
    = 99 %
  • Solubility (10%, Water):
    Pass

 

 

 :
 :307.4924 g/mol
 :GHS07
 :Ambient
 :15163-36-7
 :C₁₅H₃₃NO₃S
 :MFCD00036908