Order Entry
Austria
ContactUsLinkComponent
IPTG (isopropyl-β-D-thiogalactopyranoside) ≥99%, Ultrapure
IPTG (isopropyl-β-D-thiogalactopyranoside) ≥99%, Ultrapure
Catalog # J436-100G
Supplier:  VWR Chemicals
CAS Number:  
undefined
IPTG (isopropyl-β-D-thiogalactopyranoside) ≥99%, Ultrapure
Catalog # J436-100G
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Bottle, amber
  • Pk:
    100 g
  • Seq:
    3401
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Abs.@300 nm (5%, water):
    <0.13
  • Abs.@400 nm (5%, water):
    <0.06
  • Dioxane Test:
    Pass
  • Melting Point:
    110 – 114 °C
  • Moisture (KF):
    <1%
  • pH (5%, water) @25 °C:
    5.0 – 7.0
  • Purity:
    >99%
  • Solubility (5%, water):
    Pass
  • Specific Rotation (1%, water):
    -34.5 to -28.5°

Specification Test Results

About this item

Synonyms: IPTG, (2R,3R,4S,5R,6S)-2-(Hydroxymethyl)-6-(isopropylthio)tetrahydro-2H-pyran-3,4,5-triol, Isopropyl Beta-D-1-thiogalactopyranoside
 :
 :238.31 g/mol
Boiling Pt:402 °C (1013 hPa)
Melting Pt:109.5…110.5 °C
Storage Temperature:Freezer
EINECS:206-703-0
CAS Number:367-93-1
Formula:C₉H₁₈O₅S
MDL Number:MFCD00063273