ContactUsLinkComponent
ESD safety shoes, lace-up, TX 460
  114-4697
undefined
ESD safety shoes, lace-up, TX 460
  114-4697
  • Shoe size:
    49
  • Colour:
    Black
  • Pk:
    1 PAIR
ESD safety shoes made from high quality waterproof leather combined with safety reflectors and a MPU® Light sole (3,5 mm profile, oil, acid and petrol resistant, anti-static).
  • Skin friendly, abrasion resistant aktiv-X lining for enhanced breathability
  • Removable insoles; orthopedic insoles possible
  • 3D cushioning system: pressure relieving insole, cushioning comfort midsole, dynamic outsole system made from MPU
  • Protective steel toecap (200 J)
  • Carbon fibre reinforced toe protection
  • Electrical contact resistance of 1×10⁵ - 3,5×10⁷ Ohm
Colour: Black
 : EN 61340 (ESD); DGUV 112-191; EN ISO 20345; S2