Occupational boots, lace-up, Ranger® BGS
Supplier: HAIX SCHUHE
Certificates
About this item
Boots for every use made of 2.0 - 2.2 mm waterproof leather and Gore-Tex® lining. The rubber/PU sole with road and off-road profile has high cushioning properties, contact heat resistance and heat and cold insulation of the sole complex. It is oil and petrol resistant and antistatic.
- Waterproof and breathable thanks to Gore-Tex® lining
- Breathable, moisture is transported to the ventilation openings on the shaft and tongue
- Orthopedic properties ensure pressure relief, cushioning, correction of foot position, body straightening and muscle balance
- Sun reflection effect reduces heating of the upper leather
- Increased impact absorption and isolation in the heel area
Colour: Black
Shaft height: 17 cm
Frequently Bought Together