ESD occupational shoes, clogs, Tiefbett Leder, Munich, 11320, 11420
Deeply moulded occupational shoes made from full grain leather and an anti-slip and anti-static "A4" nitrile rubber sole; with adjustable heel strap.
- Anatomically shaped stabilising integrated insole made from cork/latex
- Insole features cushioned heel, ankle support and toe bar
- Shock absorber heel
For ESD and kitchen use.
Colour: White
Width: 9
Weight: 300 g
: EN 61340 (ESD); EN ISO 20347:2012; OB, SRC
Frequently Bought Together