Order Entry
Austria
Orders LinkContactUsLinkComponent
First Aid Box, Type 1

First Aid Box, Type 1

First aid kit, type 1, orange, for 1 - 5 people, ABS
Catalog # 118-0241
Supplier:   TRENKA
First Aid Box, Type 1
Catalog # 118-0241
Supplier:  TRENKA
First Aid Box, Type 1
First Aid Box, Type 1

First aid kit, type 1, orange, for 1 - 5 people, ABS

Price
45,60 €
Log in for availability
1 * 1 ST
Product Details & Documents

Printed ABS first aid kit for 1 to 5 workers.

  • Mobile and stationary use with wall bracket and 90° stop lock
  • Rubber seal: Dust and splash proof
  • Transparent cover plates to protect the filling parts
  • Variable interior division with dividers, sealable
  • Impact and break resistant

Color: orange

H×B×T: 200×280×115 mm

Certifications: ÖNORM Z 1020 Typ 1

Specifications
  • Description:
    First aid kit, type 1, orange, for 1 - 5 people
  • Material:
    ABS
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Komplette Nachfüllpackung für Erste-Hilfe-Koffer Typ 1 (1 - 5 Personen)
    Komplette Nachfüllpackung für Erste-Hilfe-Koffer Typ 1 (1 - 5 Personen)
    IBSA15710
    Log in for availability
    1 * 1 ST - 20,69 €