Supplier: COTTON CLASSICS HANDELS
Certificates
About this item
Specifications
- Catalog No:
- COTT02.0026GRU2XL
- COTT02.0026DGRS
- COTT02.0026BLAS
- COTT02.0026WHIM
- COTT02.0026ROTS
- COTT02.0026DGRM
- COTT02.0026WHIL
- COTT02.0026NAVM
- COTT02.0026BLAM
- COTT02.0026DGRL
- COTT02.0026WHI1XL
- COTT02.0026NAVL
- COTT02.0026BLAL
- COTT02.0026DGR1XL
- COTT02.0026WHI2XL
- COTT02.0026BLA1XL
- COTT02.0026DGR2XL
- COTT02.0026GRUS
- COTT02.0026BLA2XL
- COTT02.0026ROYS
- COTT02.0026ROTM
- COTT02.0026ROTL
- COTT02.0026ROT1XL
- COTT02.0026ROT2XL
- COTT02.0026GRUM
- COTT02.0026GRUL
- COTT02.0026NAVS
- COTT02.0026ROYM
- COTT02.0026ROYL
- COTT02.0026ROY1XL
- COTT02.0026NAV1XL
- COTT02.0026ROY2XL
- COTT02.0026GRU1XL
- COTT02.0026NAV2XL
- COTT02.0026WHIS
- Model:
- JN 26
- JN 26
- JN 26
- JN 26
- JN 26
- JN 26
- JN 26
- JN 26
- JN 26
- JN 26
- JN 26
- JN 26
- JN 26
- JN 26
- JN 26
- JN 26
- JN 26
- JN 26
- JN 26
- JN 26
- JN 26
- JN 26
- JN 26
- JN 26
- JN 26
- JN 26
- JN 26
- JN 26
- JN 26
- JN 26
- JN 26
- JN 26
- JN 26
- JN 26
- JN 26
- Garment application:
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- Gender:
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- Sleeve or leg length:
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Closure type:
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Fabric:
- 60% ring-spun cotton and 40% polyester
- 60% ring-spun cotton and 40% polyester
- 60% ring-spun cotton and 40% polyester
- 60% ring-spun cotton and 40% polyester
- 60% ring-spun cotton and 40% polyester
- 60% ring-spun cotton and 40% polyester
- 60% ring-spun cotton and 40% polyester
- 60% ring-spun cotton and 40% polyester
- 60% ring-spun cotton and 40% polyester
- 60% ring-spun cotton and 40% polyester
- 60% ring-spun cotton and 40% polyester
- 60% ring-spun cotton and 40% polyester
- 60% ring-spun cotton and 40% polyester
- 60% ring-spun cotton and 40% polyester
- 60% ring-spun cotton and 40% polyester
- 60% ring-spun cotton and 40% polyester
- 60% ring-spun cotton and 40% polyester
- 60% ring-spun cotton and 40% polyester
- 60% ring-spun cotton and 40% polyester
- 60% ring-spun cotton and 40% polyester
- 60% ring-spun cotton and 40% polyester
- 60% ring-spun cotton and 40% polyester
- 60% ring-spun cotton and 40% polyester
- 60% ring-spun cotton and 40% polyester
- 60% ring-spun cotton and 40% polyester
- 60% ring-spun cotton and 40% polyester
- 60% ring-spun cotton and 40% polyester
- 60% ring-spun cotton and 40% polyester
- 60% ring-spun cotton and 40% polyester
- 60% ring-spun cotton and 40% polyester
- 60% ring-spun cotton and 40% polyester
- 60% ring-spun cotton and 40% polyester
- 60% ring-spun cotton and 40% polyester
- 60% ring-spun cotton and 40% polyester
- 60% ring-spun cotton and 40% polyester
- Weight:
- 210 - 220 g/m²
- 210 - 220 g/m²
- 210 - 220 g/m²
- 210 - 220 g/m²
- 210 - 220 g/m²
- 210 - 220 g/m²
- 210 - 220 g/m²
- 210 - 220 g/m²
- 210 - 220 g/m²
- 210 - 220 g/m²
- 210 - 220 g/m²
- 210 - 220 g/m²
- 210 - 220 g/m²
- 210 - 220 g/m²
- 210 - 220 g/m²
- 210 - 220 g/m²
- 210 - 220 g/m²
- 210 - 220 g/m²
- 210 - 220 g/m²
- 210 - 220 g/m²
- 210 - 220 g/m²
- 210 - 220 g/m²
- 210 - 220 g/m²
- 210 - 220 g/m²
- 210 - 220 g/m²
- 210 - 220 g/m²
- 210 - 220 g/m²
- 210 - 220 g/m²
- 210 - 220 g/m²
- 210 - 220 g/m²
- 210 - 220 g/m²
- 210 - 220 g/m²
- 210 - 220 g/m²
- 210 - 220 g/m²
- 210 - 220 g/m²
Specifications
Frequently Bought Together