Disposable particle filtering respirators
Supplier: 3M
Certificates
About this item
The three-part design and the extra soft fleece on the inside of the mask ensure maximum comfort even when worn for a long time.
- Use limit: up to 4 times the limit for particles
- Straps: color yellow
- Folds flat for easy portability
- Hygienic individual packaging
- Integrated nose bridge with sweat-absorbing padding, ideal for spectacle wearers
For single use, FFP1 NR D.
Frequently Bought Together