Order Entry
Austria
ContactUsLinkComponent
Rubber stoppers for lyophilisation ampoules, Vacule®, WHEATON®
Rubber stoppers for lyophilisation ampoules, Vacule®, WHEATON®
  WHEAW651450
undefined
Rubber stoppers for lyophilisation ampoules, Vacule®, WHEATON®
  WHEAW651450

 

  • Ø bottom:
    7,0 mm
  • Ø top:
    13,0 mm
  • Description:
    For 2 ml lyophilisation ampoules
  • Environmentally Preferable:
  • Pk:
    144

 

 

These stoppers are made from grey butyl rubber.