Order Entry
Austria
Orders LinkContactUsLinkComponent
Probes for 112

Probes for 112

Supplier:   Testo
Probes for 112
Supplier:  Testo
Certificates
Probes for 112

9 Options Available Below

Product Details & Documents
Product Family Options

Product Information

  • Range
    Accuracy
    Application
    Length
    Ø×L
    Availability
    Price
  • 662-3143
    Stainless steel probe with PUR cable
    Stainless steel probe with PUR cable
    −50...+150 °C
    ±0,5% of mv (100...150 °C)
    ±0,2 °C (−25 ...+74,9 °C)
    ±0,4 °C (remaining range)
    Food
    125 mm
    4×125 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 123,00 €
    123,00 €
    1 * 1 ST
       123,00 €

  • 620-3660
    Robust probe with special handle, with PUR cable
    Robust probe with special handle, with PUR cable
    −25...+150 °C
    ±0,5% of mv (100...150 °C)
    ±0,2 °C (−25...+74,9 °C)
    ±0,4 °C (remaining range)
    Food
    115 mm
    5×115 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 117,00 €
    117,00 €
    1 * 1 ST
       117,00 €

  • 628-3306
    NTC probe, corkscrew design
    NTC probe, corkscrew design
    −50...+140 °C
    ±0,5% of mv (100...140 °C)
    ±0,2 °C (−25...+74,9 °C)
    ±0,4 °C (remaining range)
    Frozen food
    110 mm
    8×110 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 186,00 €
    186,00 €
    1 * 1 ST
       186,00 €

  • 620-3233
    Pipe wrap probe, with Velcro®
    Pipe wrap probe, with Velcro®
    −50...+70 °C
    ±0,2 °C (−25...+70 °C)
    ±0,4 °C (−50...−25,1 °C)
    Surface
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 100,00 €
    100,00 €
    1 * 1 ST
       100,00 €

  • 620-3316
    Waterproof NTC probe, calibratable
    Waterproof NTC probe, calibratable
    −25...+120 °C
    ±0,5% of mv (100...120 °C)
    ±0,2 °C (−25...+74,9 °C)
    ±0,4 °C (remaining range)
    Immersion/penetration
    160 mm
    5×160 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 87,00 €
    87,00 €
    1 * 1 ST
       87,00 €

  • TEST0614.1272
    Waterproof PT 100 probe, calibratable
    Waterproof PT 100 probe, calibratable
    −50...+300 °C
    Class A
    Immersion/penetration
    160 mm
    5×160 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 95,00 €
    95,00 €
    1 * 1 ST
       95,00 €

  • TEST0614.1712
    NTC probe, robust, accurate
    NTC probe, robust, accurate
    −25...+120 °C
    ±0,5% of mv (100...120 °C)
    ±0,2 °C (−25...+74,9 °C)
    ±0,4 °C (remaining range)
    Air
    115 mm
    5×115 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 90,00 €
    90,00 €
    1 * 1 ST
       90,00 €

  • 620-3235
    NTC probe, stainless steel, with PUR cable
    NTC probe, stainless steel, with PUR cable
    −25...+120 °C
    ±0,5% of mv (100...120 °C)
    ±0,2 °C (−25...+74,9 °C)
    ±0,4 °C (remaining range)
    Food
    125 mm
    4×125 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 133,00 €
    133,00 €
    1 * 1 ST
       133,00 €

  • TEST0614.2272
    PT100 probe, calibratable
    PT100 probe, calibratable
    −50...+300 °C
    Class A
    Food
    125 mm
    4×125 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 112,00 €
    112,00 €
    1 * 1 ST
       112,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.