ContactUsLinkComponent
Mini Tubes, SP Bel-Art
Mini Tubes, SP Bel-Art
Catalog # 391-9030
undefined
Mini Tubes, SP Bel-Art
Catalog # 391-9030
Specifications
  • Capacity:
    1 ml
  • Description:
    400×12 tube strips
  • Pk:
    4.800
Specifications
About this item
Made of virgin, medical grade polypropylene.
  • Autoclavable
  • Frozen to −20 °C
  • Very good chemical resistance
Mini-Tubes offer a convenient and economical means of performing laboratory procedures using multi-channel pipettors of four, eight or twelve channels. Features an attachment bridge between tubes, forming a Mini-Tube strip; can be easily separated into single tubes by cutting apart.