Accessories for multi-parameter meter (pH/Conductivity/Oxygen), handheld, SensoDirect 150
Supplier: Lovibond
Certificates
Accessories for multi-parameter meter (pH/Conductivity/Oxygen), handheld, SensoDirect 150
Supplier: Lovibond
About this item
Frequently Bought Together
VWR® B'Wipes 6000, Cleanroom wipes
Supplier: VWR Collection
Wipes for general applications, Lavette Super
Supplier: PGI NONWOVEN
VWR® Spec-Wipe® 3, Cleanroom Wipes, Non Woven, Sterile
Supplier: VWR Collection
Nitrile gloves, TouchNTuff® 92-600 / 92-605
Supplier: Ansell
pH electrodes, InLab®, Expert Series
Supplier: Mettler - Toledo
VWR®, Syringe Filters
Supplier: VWR Collection
Single use syringes, 2-piece, Injekt® Solo
Supplier: B. Braun
Colour Lab Tapes
Supplier: GA
BD Discardit™ II Syringes
Supplier: BD Medical
pH electrodes for fast results, InLab®
Supplier: Mettler - Toledo
VWR® Spec-Wipe® 3, Cleanroom Wipes, Pre-Saturated
Supplier: VWR Collection
VWR® Facial Tissues
Supplier: VWR Collection
VWR®, Cleanroom Wipes, 100% Polyester
Supplier: VWR Collection
Cleaning tools, EasyReach™
Supplier: CONTEC
VWR®, Nitrile Gloves
Supplier: VWR Collection
Folded towels, zigzag fold, ZetForm®
Supplier: Zellstoff-Vertriebs-GmbH & Co. KG
Facial tissues, KLEENEX®
Supplier: KIMBERLY CLARK
Conductivity meter, handheld, SensoDirect Con110
Supplier: Lovibond