Order Entry
Austria
Orders LinkContactUsLinkComponent
 

pH/°C testers, 206

Supplier:   Testo
pH/°C testers, 206
Supplier:  Testo
Certificates
pH/°C testers, 206

5 Options Available Below

Product Details & Documents

The testo 206 series is ideal for pH measurements in liquid and semi-solid media. The instruments offer a combination of a pH immersion probe and a temperature probe for precise and fast temperature compensation. Thanks to the large volume of gel electrolyte and the dual wall diaphragm, the probe is leak-proof, maintenance-free and insensitive to dirt.

  • Built-in temperature sensor
  • Maintenance-free gel electrolyte
  • Robust, waterproof and dishwasher-safe protective cover (TopSafe, protection class IP 65)
  • Automatic final value recognition (Auto-hold)
  • 1, 2, or 3-point calibration possible

Testo 206-pH1 is specially designed for fast pH checks in liquids.

Testo 206-pH2 is ideally suited for pH measurement in semi-solid and protein-rich foods such as jellies, creams, cheese, fruit etc.

Testo 206-pH3 is equipped with a BNC socket which allows the connection of any pH probes to the instrument, depending on the application. This makes the instrument universally applicable.

 : Single instruments are supplied with TopSafe cover and belt/wall holder. pH1 and pH2 also include a storage cap with gel.
pH1 and pH2 starter kits are supplied with storage cap with gel, calibration dosing bottles 250 ml pH 4 and 7, TopSafe cover, belt/wall holder and aluminium case.
All instruments are supplied with probe head but without probe. Probes need to be ordered separately.

Specifications for Product Family

Specifications

  • Catalog No:
  • TEST0563.2061
  • TEST0563.2065
  • TEST0563.2062
  • TEST0563.2066
  • TEST0563.2063
  • Model:
  • 206 pH1
  • 206 pH1
  • 206 pH2
  • 206 pH2
  • 206 pH3
  • pH range:
  • 0...+14
  • 0...+14
  • 0...+14
  • 0...+14
  • 0...+14
  • pH resolution:
  • 0,01
  • 0,01
  • 0,01
  • 0,01
  • 0,01
  • pH accuracy:
  • ±0,02
  • ±0,02
  • ±0,02
  • ±0,02
  • ±0,02
  • pH calibration:
  • 1, 2, or 3-point calibration
  • 1, 2, or 3-point calibration
  • 1, 2, or 3-point calibration
  • 1, 2, or 3-point calibration
  • 1, 2, or 3-point calibration
  • Temperature range:
  • 0...+60 °C
  • 0...+60 °C
  • 0...+60 °C
  • 0...+60 °C
  • Depending on sensor
  • Temperature resolution:
  • 0,1 °C
  • 0,1 °C
  • 0,1 °C
  • 0,1 °C
  • 0,1 °C
  • Temperature accuracy:
  • ±0,4 °C
  • ±0,4 °C
  • ±0,4 °C
  • ±0,4 °C
  • ±0,4 °C
  • Temperature compensation:
  • Automatic
  • Automatic
  • Automatic
  • Automatic
  • Automatic
  • Display:
  • LCD, 2 lines
  • LCD, 2 lines
  • LCD, 2 lines
  • LCD, 2 lines
  • LCD, 2 lines
  • Power supply:
  • 1 × CR2032 battery
  • 1 × CR2032 battery
  • 1 × CR2032 battery
  • 1 × CR2032 battery
  • 1 × CR2032 battery
  • Compliance:
  • IP68
  • IP68
  • IP68
  • IP68
  • IP68
  • W×D×H:
  • 33×20×197 mm
  • 33×20×197 mm
  • 33×20×197 mm
  • 33×20×197 mm
  • 33×20×197 mm
  • Weight:
  • 0,069 kg
  • 0,069 kg
  • 0,062 kg
  • 0,062 kg
  • 0,062 kg

Specifications

Product Family Options

pH1

Product Information

  • Availability
    Price
  • TEST0563.2061
    pH/°C tester, 206 pH1, single instrument
    pH/°C tester, 206 pH1, single instrument
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 155,00 €
    155,00 €
    1 * 1 ST
       155,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    TEST0563.2061
    Model
    206 pH1
    pH range
    0...+14
    pH resolution
    0,01
    pH accuracy
    ±0,02
    pH calibration
    1, 2, or 3-point calibration
    Temperature range
    0...+60 °C
    Temperature resolution
    0,1 °C
    Temperature accuracy
    ±0,4 °C
    Temperature compensation
    Automatic
    Display
    LCD, 2 lines
    Power supply
    1 × CR2032 battery
    Compliance
    IP68
    W×D×H
    33×20×197 mm
    Weight
    0,069 kg
  • TEST0563.2065
    pH/°C tester, 206 pH1, starter kit
    pH/°C tester, 206 pH1, starter kit
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 259,00 €
    259,00 €
    1 * 1 ST
       259,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    TEST0563.2065
    Model
    206 pH1
    pH range
    0...+14
    pH resolution
    0,01
    pH accuracy
    ±0,02
    pH calibration
    1, 2, or 3-point calibration
    Temperature range
    0...+60 °C
    Temperature resolution
    0,1 °C
    Temperature accuracy
    ±0,4 °C
    Temperature compensation
    Automatic
    Display
    LCD, 2 lines
    Power supply
    1 × CR2032 battery
    Compliance
    IP68
    W×D×H
    33×20×197 mm
    Weight
    0,069 kg

pH2

Product Information

  • Availability
    Price
  • TEST0563.2062
    pH/°C tester, 206 pH2, single instrument
    pH/°C tester, 206 pH2, single instrument
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 259,00 €
    259,00 €
    1 * 1 ST
       259,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    TEST0563.2062
    Model
    206 pH2
    pH range
    0...+14
    pH resolution
    0,01
    pH accuracy
    ±0,02
    pH calibration
    1, 2, or 3-point calibration
    Temperature range
    0...+60 °C
    Temperature resolution
    0,1 °C
    Temperature accuracy
    ±0,4 °C
    Temperature compensation
    Automatic
    Display
    LCD, 2 lines
    Power supply
    1 × CR2032 battery
    Compliance
    IP68
    W×D×H
    33×20×197 mm
    Weight
    0,062 kg
  • TEST0563.2066
    pH/°C tester, 206 pH2, starter kit
    pH/°C tester, 206 pH2, starter kit
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 355,00 €
    355,00 €
    1 * 1 ST
       355,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    TEST0563.2066
    Model
    206 pH2
    pH range
    0...+14
    pH resolution
    0,01
    pH accuracy
    ±0,02
    pH calibration
    1, 2, or 3-point calibration
    Temperature range
    0...+60 °C
    Temperature resolution
    0,1 °C
    Temperature accuracy
    ±0,4 °C
    Temperature compensation
    Automatic
    Display
    LCD, 2 lines
    Power supply
    1 × CR2032 battery
    Compliance
    IP68
    W×D×H
    33×20×197 mm
    Weight
    0,062 kg

pH3

Product Information

  • Availability
    Price
  • TEST0563.2063
    pH/°C tester, 206 pH3, single instrument
    pH/°C tester, 206 pH3, single instrument
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 127,00 €
    127,00 €
    1 * 1 ST
       127,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    TEST0563.2063
    Model
    206 pH3
    pH range
    0...+14
    pH resolution
    0,01
    pH accuracy
    ±0,02
    pH calibration
    1, 2, or 3-point calibration
    Temperature range
    Depending on sensor
    Temperature resolution
    0,1 °C
    Temperature accuracy
    ±0,4 °C
    Temperature compensation
    Automatic
    Display
    LCD, 2 lines
    Power supply
    1 × CR2032 battery
    Compliance
    IP68
    W×D×H
    33×20×197 mm
    Weight
    0,062 kg

Probes

Product Information

  • Catalog #
    For
    Diaphr.
    Ø×L
    pH
    Electrolyte
    Description
    Connection
    Availability
    Price
  • Food
    Hole
    6,5×95 mm
    2 - 14
    Gel
    pH penetration electrode, unbreakable plastic coating around glass electrode, incl. watering cap, 124 g
    BNC, fixed cable
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 261,00 €
    261,00 €
    1 * 1 ST
       261,00 €

Product Information

  • Catalog #
    For
    Connection
    Diaphr.
    Ø×L
    pH
    Description
    Electrolyte
    Availability
    Price
  • General purposes
    0...80 °C
    01
    12×120 mm
    0 - 14
    pH probe, glass, for testo 206 pH3, with temperature sensor
    Gel
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 265,00 €
    265,00 €
    1 * 1 ST
       265,00 €

Product Information

  • Catalog #
    For
    Connection
    Diaphr.
    Ø×L
    pH
    Description
    Electrolyte
    Availability
    Price
  • Liquids
    0...60 °C
    02
    15×3,5 mm
    0 - 14
    Spare pH probe for testo 206 pH1, with NTC, incl. gel storage cap
    Gel
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 88,00 €
    88,00 €
    1 * 1 ST
       88,00 €

Product Information

  • Catalog #
    For
    Connection
    Diaphr.
    Ø×L
    pH
    Description
    Electrolyte
    Availability
    Price
  • Semi-solid substances
    0...60 °C
    03
    11×35 mm
    0 - 14
    Spare pH probe for testo 206 pH2, with NTC, incl. gel storage cap
    Gel
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 177,00 €
    177,00 €
    1 * 1 ST
       177,00 €

Product Information

  • Catalog #
    For
    Connection
    Diaphr.
    Ø×L
    pH
    Description
    Electrolyte
    Availability
    Price
  • General purposes
    - °C
    05
    12×120 mm
    0 - 14
    Universal pH probe, plastic, for testo 206 pH3, without temperature sensor
    Gel
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 116,00 €
    116,00 €
    1 * 1 ST
       116,00 €

Product Information

  • Catalog #
    For
    Connection
    Diaphr.
    Ø×L
    pH
    Description
    Electrolyte
    Availability
    Price
  • General purposes
    0...80 °C
    04
    12×120 mm
    0 - 14
    Universal pH probe, plastic, for testo 206 pH3, with temperature sensor
    Gel
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 224,00 €
    224,00 €
    1 * 1 ST
       224,00 €

Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Storage cap for testo 206, with KCI gel filling
    Storage cap for testo 206, with KCI gel filling
    TEST0554.2067
    Log in for availability
    1 * 1 ST - 17,00 €

Product Information

  • Catalog #
    Availability
    Price
  • pH buffer solution 7,00 in dosing bottle (250 ml), with DAkkS calibration certificate
    pH buffer solution 7,00 in dosing bottle (250 ml), with DAkkS calibration certificate
    TEST0554.2063
    Log in for availability
    1 * 250 mL - 21,00 €

Product Information

  • Catalog #
    Availability
    Price
  • pH buffer solution 4,01 in dosing bottle (250 ml), with DAkkS calibration certificate
    pH buffer solution 4,01 in dosing bottle (250 ml), with DAkkS calibration certificate
    TEST0554.2061
    Log in for availability
    1 * 250 mL - 21,00 €