Surgical gowns, basic and high protection
These fluid-repellent, latex-free gowns are all made of non woven SMMS fabric. They provide protection from low, medium or high fluid exposure.
Basic:
- For procedures with low levels of fluid
- Good strength characteristics
- Low linting
- Ultrasonically bonded seams
- Overlapping back
- Adjustable neckline
High protection:
- For medium (standard) to high (reinforced) fluid exposure
- Alcohol repellent
- Good strength characteristics
- Low linting
- Ultrasonically bonded seams
- Overlapping back
- Lightweight and comfortable
- Adjustable neckline
- Larger sizes have a longer length
: EN 13795