Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Homogenisers
Dispersing Tools
VWR®, Containers and blades for Sterilmixer 12
VWR®, Containers and blades for Sterilmixer 12
VWR®, Containers and blades for Sterilmixer 12
Catalog # 710-0953
Supplier:
VWR Collection
Expand image
VWR®, Containers and blades for Sterilmixer 12
Catalog # 710-0953
Supplier:
VWR Collection
Specifications
Description:
Johnny sealing ring for blades, autoclavable, for use with Steri-Blostev 12 base fixing plate
Seq:
05
Pk:
1
Specifications
About this item
Product Details
Product Details