Order Entry
Austria
ContactUsLinkComponent
Dragendorff's reagent, spray reagent for TLC, Supelco®
Flammable
Corrosive
Irritant
Dragendorff's reagent, spray reagent for TLC, Supelco®
Catalog # 1.02035.0100
Supplier:  Merck
CAS Number:  
Flammable
Corrosive
Irritant
Dragendorff's reagent, spray reagent for TLC, Supelco®
Catalog # 1.02035.0100
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 ml
  • Seq:
    2440

Specifications

Specification Test Results

  • Boiling Point:
    77 °C
  • Density:
    0.95 g/cm3 (20 °C)
  • Hazard Class:
    3
  • Hazard Class Description:
    Flammable Liquid

Specification Test Results

About this item

 :
Boiling Pt:77 °C (1013 hPa)
Density:1.2 g/cm³ (20 °C)
UN:2924
ADR:3 (8),II
 :M-Clarity™ quality level = MQ100
Flash Pt:–4 °C
 :GHS02
 :GHS05
 :GHS07
Storage Temperature:Ambient
CAS Number:39775-75-2
MDL Number:MFCD00283106