VWR®, Bovine Ribonuclease A (from Pancreas)
Ribonuclease A is an endoribonuclease that efficiently hydrolyses RNA from tissue or bacterial cell cultures.
- Useful during DNA purification
- High purity
- Available in a ready-to-use solution
RNase A is used for a variety of molecular biology applications, most commonly for the hydrolysis of RNA that contaminates DNA preparations. This enzyme is used during plasmid purification without nicking or degrading plasmid.
- CAS No.:9001-99-4
- Source:Pancreas
- Species:Bovine
- Storage conditions:Refrigerator
- Form:solution
- Enzyme name:Ribonuclease A
- Enzyme synonyms:RNase A
- Concentration:10 mg/ml
- pH:pH 8
- Grade:For biotechnology
Frequently Bought Together