Bovine Ribonuclease A (from Pancreas)
Supplier: PanReac AppliChem
Certificates
About this item
DNAse-free
Specifications
- CAS No.:9001-99-4
- Source:Pancreas
- Species:Bovine
- Storage conditions:Freezer
- MDL No.:MFCD00132184
- Enzyme name:Ribonuclease A
- Enzyme activity:90 U/mg (Kunitz)
- Enzyme synonyms:RNase A
Specifications
Frequently Bought Together