Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Buffer, TRIS-glycine solution 10X concentrate (TG buffer), Sigma-Aldrich®

TRIS-glycine buffer solution 10X concentrate (TG buffer), Glass bottle, 1 L
Catalog # SIAL93321-1L
Supplier:   Merck
Buffer, TRIS-glycine solution 10X concentrate (TG buffer), Sigma-Aldrich®
Catalog # SIAL93321-1L
Supplier:  Merck
CAS Number:  
Buffer, TRIS-glycine solution 10X concentrate (TG buffer), Sigma-Aldrich®

TRIS-glycine buffer solution 10X concentrate (TG buffer), Glass bottle, 1 L

CAS Number:  
Price
143,00 €
Log in for availability
1 * 1 L
Product Details & Documents
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Description:
    TRIS-glycine buffer solution 10X concentrate (TG buffer)
  • Cat. No.:
    SIAL93321-1L
  • M-Clarity quality level:
    MQ100

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.