Order Entry
Austria
Orders LinkContactUsLinkComponent
Buffer, TE solution (TRIS-EDTA buffer) pH 8,0

Buffer, TE solution (TRIS-EDTA buffer) pH 8,0

TE buffer solution (TRIS-EDTA buffer) pH 8,0, Glass bottle, 1 L
Catalog # ROCKMB-040
Buffer, TE solution (TRIS-EDTA buffer) pH 8,0
Catalog # ROCKMB-040
CAS Number:  
Buffer, TE solution (TRIS-EDTA buffer) pH 8,0
Buffer, TE solution (TRIS-EDTA buffer) pH 8,0

TE buffer solution (TRIS-EDTA buffer) pH 8,0, Glass bottle, 1 L

CAS Number:  
Price
199,00 €
Log in for availability
1 * 1 L
Product Details & Documents

Documents

Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Description:
    TE buffer solution (TRIS-EDTA buffer) pH 8,0

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.