Buffer, SSC solution 20X concentrate (saline-sodium citrate), Corning®
Artikelnr. MDTC46-020-CM
Lieferant: Corning
CAS-Nummer:
Spezifikationen
- Pack type:Glass bottle
- Pk:6
- Description:SSC buffer solution 20X concentrate (saline-sodium citrate)
Spezifikationen
Über diesen Artikel
20X Saline-Sodium Citrate (SSC) buffer is used in both nucleic acid hybridization and blot transfer procedures
20X is the maximum concentration used; 0.2X may be used depending on the application.