Order Entry
Austria
ContactUsLinkComponent
Buffer, SSC solution 20X concentrate (saline-sodium citrate)
Buffer, SSC solution 20X concentrate (saline-sodium citrate)
Catalog # 733-1635
Supplier:  LONZA
CAS Number:  
Buffer, SSC solution 20X concentrate (saline-sodium citrate)
Catalog # 733-1635
Supplier:  LONZA
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Description:
    SSC buffer solution 20X concentrate (saline-sodium citrate)

Specifications

About this item