Order Entry
Austria
ContactUsLinkComponent
Buffer, MOPS solution 10X concentrate
Buffer, MOPS solution 10X concentrate
Catalog # 733-1628
Supplier:  LONZA
CAS Number:  
Buffer, MOPS solution 10X concentrate
Catalog # 733-1628
Supplier:  LONZA
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Description:
    MOPS buffer solution 10X concentrate

Specifications

About this item