Order Entry
Austria
Orders LinkContactUsLinkComponent
Buffer, EDTA buffer solution pH 8,0 (0,5 mol/l)

Buffer, EDTA buffer solution pH 8,0 (0,5 mol/l)

EDTA buffer solution pH 8,0 (0,5 mol/l), 1 KIT
Catalog # R1021
Buffer, EDTA buffer solution pH 8,0 (0,5 mol/l)
Catalog # R1021
CAS Number:  
Buffer, EDTA buffer solution pH 8,0 (0,5 mol/l)
Buffer, EDTA buffer solution pH 8,0 (0,5 mol/l)

EDTA buffer solution pH 8,0 (0,5 mol/l), 1 KIT

CAS Number:  
Price
27,60 €
Log in for availability
1 * 1 KIT
Product Details & Documents
Specifications

Specifications

  • Pk:
    1 KIT
  • Description:
    EDTA buffer solution pH 8,0 (0,5 mol/l)

Specifications