Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Buffers
Cloning Buffers
Buffer, EDTA buffer solution pH 8,0 (0,5 mol/l)
Buffer, EDTA buffer solution pH 8,0 (0,5 mol/l)
Buffer, EDTA buffer solution pH 8,0 (0,5 mol/l)
Catalog # APPCA3145.1000
Supplier:
PanReac AppliChem
CAS Number:
Expand image
Buffer, EDTA buffer solution pH 8,0 (0,5 mol/l)
Catalog # APPCA3145.1000
Supplier:
PanReac AppliChem
CAS Number:
Specifications
Pack type:
Glass bottle
Pk:
1 L
Description:
EDTA buffer solution pH 8,0 (0,5 mol/l)
Specifications
About this item
Product Details
Product Details