Order Entry
Austria
ContactUsLinkComponent
Buffer, PBS solution (phosphate buffered saline) pH 7,4, Sigma-Aldrich®
Buffer, PBS solution (phosphate buffered saline) pH 7,4, Sigma-Aldrich®
Catalog # SIALP5368-10PAK
Supplier:  Merck
CAS Number:  
Buffer, PBS solution (phosphate buffered saline) pH 7,4, Sigma-Aldrich®
Catalog # SIALP5368-10PAK
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    10
  • Description:
    PBS buffer solution (phosphate buffered saline) pH 7,4
  • M-Clarity quality level:
    MQ200

Specifications

About this item