Buffer, PBS solution 10X concentrate (phosphate buffered saline), MP Biomedicals
Supplier: MP Biomedicals
Certificates
Buffer, PBS solution 10X concentrate (phosphate buffered saline), MP Biomedicals
Supplier: MP Biomedicals
About this item
Frequently Bought Together
Ethyl acetate anhydrous, 99.8 % 1 * 2 L
Supplier: Merck
2,5-Di-tert-butylhydroquinone for synthesis, Sigma-Aldrich®
Supplier: Merck
Methanol ≥99.9% (by GC) Reag. Ph. Eur., ACS, ISO for analysis
Supplier: PanReac AppliChem
Chlorobenzene ≥99%
Supplier: Thermo Fisher Scientific
Dichloromethane 99.8% stabilised for HPLC
Supplier: Thermo Fisher Scientific
N,N-Dimethylacetamide 99%, pure
Supplier: Thermo Fisher Scientific
Aeroflow tap, na, general spare part 1 * 1 items
Supplier: Avantor
Petroleum spirit, 80…110 °C
Supplier: Roth Carl
Dibutyl ether, extra dry 99+%, AcroSeal®
Supplier: Thermo Fisher Scientific
METHANOL SUITABLE FOR HPLC >99.9% 1 * 2,5 L
Supplier: Merck
Ethyl acetate ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
N,N-Dimethylformamide ≥99.8% ACS for spectroscopy
Supplier: Thermo Fisher Scientific
Toluene 99.5% ACS for spectroscopy
Supplier: Thermo Fisher Scientific
tert-Butyl methyl ether, extra dry over Molecular Sieve 99+%, AcroSeal®
Supplier: Thermo Fisher Scientific
N,N-Dimethylformamide ≥99.7% for HPLC
Supplier: Thermo Fisher Scientific