Order Entry
Austria
Orders LinkContactUsLinkComponent
VWR® UltraPure, Buffer, PBS solution 10X concentrate (phosphate buffered saline)

VWR® UltraPure, Buffer, PBS solution 10X concentrate (phosphate buffered saline)

PBS buffer solution 10X concentrate (phosphate buffered saline), Plastic bottle, 500 mL
Catalog # K813-500ML
Supplier:   VWR Chemicals
VWR® UltraPure, Buffer, PBS solution 10X concentrate (phosphate buffered saline)
Catalog # K813-500ML
Supplier:  VWR Chemicals
CAS Number:  
VWR® UltraPure, Buffer, PBS solution 10X concentrate (phosphate buffered saline)
VWR® UltraPure, Buffer, PBS solution 10X concentrate (phosphate buffered saline)

PBS buffer solution 10X concentrate (phosphate buffered saline), Plastic bottle, 500 mL

CAS Number:  
Price
113,00 €
Log in for availability
1 * 500 mL
Product Details & Documents
  • No DNases, RNases or proteases detected

pH @ 25 °C (1:10 water)    6,6 to 7,2

Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Description:
    PBS buffer solution 10X concentrate (phosphate buffered saline)

Specifications