Dextran sulphate sodium salt, M.W. ~ 40,000
Catalog # A3261.0250
Supplier: PanReac AppliChem
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:250 g
- Seq:3545
- Environmentally Preferable:
Specifications
About this item
Synonyms: Dextran sulphate sodium from Dextran
:
CAS Number:9011-18-1
Formula:(C₆H₇Na₃O₁₄S₃)n
MDL Number:MFCD00081551