Dextran sulphate sodium salt, Calbiochem®
Supplier: Merck Millipore (Calbiochem)
Certificates
About this item
Synonyms: Dextran sulphate sodium from Dextran
CAS Number:9011-18-1
:
Formula:(C₆H₇Na₃O₁₄S₃)n
MDL Number:MFCD00081551
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together