Order Entry
Austria
ContactUsLinkComponent
Phenol ≥99% saturated with buffer for biotechnology, buffered
Irritant
Corrosive
Health
Phenol ≥99% saturated with buffer for biotechnology, buffered
Supplier:  VWR Chemicals
Certificates
undefined
Irritant
Corrosive
Health
Phenol ≥99% saturated with buffer for biotechnology, buffered
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Abs.@ 330nm:
    = 0.2
  • Abs.@ 405nm:
    = 0.1
  • Abs.@ 510nm:
    = 0.1
  • pH (Buffer) @25°C:
    7.7 - 8.1
  • Purity:
    = 99 %

Specification Test Results

Related Information

Frequently Bought Together