Order Entry
Austria
ContactUsLinkComponent
Buffer, TAE solution 10X concentrate (TRIS-acetate-EDTA buffer), Sigma-Aldrich®
Buffer, TAE solution 10X concentrate (TRIS-acetate-EDTA buffer), Sigma-Aldrich®
Catalog # SIALT8280-1L
Supplier:  Merck
CAS Number:  
Buffer, TAE solution 10X concentrate (TRIS-acetate-EDTA buffer), Sigma-Aldrich®
Catalog # SIALT8280-1L
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Description:
    TAE buffer solution 10X concentrate (TRIS-acetate-EDTA buffer)
  • M-Clarity quality level:
    MQ200

Specifications

About this item