ContactUsLinkComponent
Microplate tilting stand, SP Bel-Art
Microplate tilting stand, SP Bel-Art
  BELAF189380000
undefined
Microplate tilting stand, SP Bel-Art
  BELAF189380000
  • Description:
    Microplate tilting stand, 114×165×57 mm
  • Pk:
    1
Accurate, repetitive aliquoting into microplate wells is simplified by placing the plate on the adjustable stand and setting the slant to an optimal angle. The slanted plate permits the user to track which wells have received aliquots by visibly contrasting the menisci between adjoining wells.
  • Promotes addition of accurate aliquots to microplate wells
  • Effortlessly view additions at any angle
  • Reduces operator error during pipetting