Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
TLC Plates
TLC plates
TLC plates
TLC plates
Catalog # 552-0129
Supplier:
GERBER FUNKE
Expand image
TLC plates
Catalog # 552-0129
Supplier:
GERBER FUNKE
Specifications
Coating material:
Silica gel
Plate size:
200×200 mm
Seq:
01
Pk:
25
Specifications
About this item
Product Details
With aluminium liner
Can be cut with scissors
Product Details