ContactUsLinkComponent
Microcentrifuge tube opener, SP Bel-Art
Microcentrifuge tube opener, SP Bel-Art
Catalog # BELAF199250001
undefined
Microcentrifuge tube opener, SP Bel-Art
Catalog # BELAF199250001
Specifications
  • Description:
    Microcentrifuge tube opener
  • Pk:
    3
Specifications
About this item
Easily open all sizes of microcentrifuge tubes with this double-ended tool.
  • Opens standard 1,5, 2,0 and 2,2 ml tubes with one end
  • Forked blade on other end opens smaller sizes (250, 400 and 550 μl)
  • Dimensions: 7 cm (3⅛") long