Orders LinkContactUsLinkComponent
Ball valves with thread
Ball valves with thread
Supplier:   BURKLE
Ball valves with thread
Supplier:  BURKLE
Certificates
Ball valves with thread
Ball valves with thread
5 Options Available Below
Product Details & Documents
Ball valve made of PP with outer/inner thread.
  • Maximum operating pressure 10 bar (at +20 °C)
  • Maximum operating temperature 100 °C
  • Suitable for food
It also consists fibre-glass reinforced, ball made of PP, handle with O-ring made of food-safe NBR and a seal made of HDPE.
Product Family Options
Product Information
  • Hole Ø
    Thread
    Availability
    Price
  • 15 mm
    12,7 mm inner inflowing/M29×1,75 outer outflowing
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 20,00 €
    20,00 €
    1 * 1 ST
       20,00 €
  • 20 mm
    19,05 mm inner inflowing/M36×3 outer outflowing
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 23,90 €
    23,90 €
    1 * 1 ST
       23,90 €
  • 20 mm
    19,05 mm outer inflowing/M36×3 outer outflowing
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 26,00 €
    26,00 €
    1 * 1 ST
       26,00 €
  • 15 mm
    12,7 mm outer inflowing/M29×1,75 outer outflowing
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 22,30 €
    22,30 €
    1 * 1 ST
       22,30 €
  • 25 mm
    25,4 mm outer inflowing/M42×3 outer outflowing
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 31,60 €
    31,60 €
    1 * 1 ST
       31,60 €