ContactUsLinkComponent
Tubing connector assortment, SP Bel-Art
Tubing connector assortment, SP Bel-Art
Catalog # 241-2271
undefined
Tubing connector assortment, SP Bel-Art
Catalog # 241-2271
Specifications
  • Pk:
    1 SET
Specifications
About this item
Complete fitting assortment box with connectors for tubings.
  • Assortment of 74 connector fittings
  • Clear, hinge-top, polystyrene storage box
Ordering information: Kits consists of polypropylene tubing connectors 5 pairs, straight stepped tubing connectors 6 pairs, "T" connectors 5 pairs, "Y" connectors 6 pairs and polyethylene quick disconnects 3 pairs, cup-type 12 pairs.