Tubing connectors, '5 in 1', SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
Certificates
About this item
PP
- Fits 5 sizes of tubing and will connect the same size tubing on both ends, or will act as a reducer for any combination within the range
- Inside diameter increases with notch size when unneeded length is cut off for unrestricted flow
Frequently Bought Together