Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Seals
Sealing Rings
Filter seal, Vac-Ring™, SP Bel-Art
Filter seal, Vac-Ring™, SP Bel-Art
BELAH389590000
:
Bel-Art Products, a Part of SP
Filter seal, Vac-Ring™, SP Bel-Art
BELAH389590000
:
Bel-Art Products, a Part of SP
Description:
Filter seal, Vac-Ring™, black, for funnel stems up to 22 mm Ø, thickness 9,5 mm and O.Ø 76 mm
Material:
Neoprene
Pk:
1