ContactUsLinkComponent
Slide holder, SP Bel-Art
Slide holder, SP Bel-Art
Catalog # 631-9109
Slide holder, SP Bel-Art
Catalog # 631-9109
Specifications
  • No. of slides:
    12
  • Description:
    Slide holders
  • Material:
    PS
  • W×D×H:
    86×178×19 mm
  • Pk:
    6
Specifications
About this item
Polystyrene holder for up to 12 microscope slides designed for preparation or staining.
  • Inclined ribs for holding slides on their ends
  • Robust and durable white plastic design
  • Safe storage with visible hinged cases
Packaging: Sold by six items.