Slide tray and cabinet, SP Bel-Art
This tray and cabinet system can hold up to 240 slides in a dust-free enclosure.
- Cabinet and trays sold separately
- White PS background to distinguish dyes
- Handy finger cut out simplifies removal of slides
: Includes PP outer shell, PS tray rails and a vertically sliding, clear acrylic door with a knob for easy opening.
Frequently Bought Together